Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diethylmethylamine 98.0+%, TCI America™
SDP

Catalog No. D1138500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1138500ML 500 mL
D113825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1138500ML Supplier TCI America Supplier No. D1138500ML

Chemical Identifiers

CAS 616-39-7
Molecular Formula C5H14N
Molecular Weight (g/mol) 88.17
MDL Number MFCD00009049
InChI Key GNVRJGIVDSQCOP-UHFFFAOYSA-O
Synonym n,n-diethylmethylamine, diethylmethylamine, ethanamine, n-ethyl-n-methyl, n-methyldiethylamine, diethylamine, n-methyl, n-ethyl-n-methyl-ethanamine, methyldiethylamine, diethylmethyl amine, diethyl methyl amine, methyl diethyl amine
PubChem CID 12022
IUPAC Name diethyl(methyl)azanium
SMILES CC[NH+](C)CC

Specifications

Color Yellow
Boiling Point 62°C
Quantity 500 mL
UN Number 2733
Formula Weight 87.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Diethylmethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.