Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diethylnicotinamide, 97%

Catalog No. p-110109
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

CAS: 59-26-7 | C10H14N2O | 178.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-26-7
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.23
MDL Number MFCD00006386
InChI Key NCYVXEGFNDZQCU-UHFFFAOYSA-N
Synonym nikethamide, n,n-diethylnicotinamide, nicethamide, corvin, anacardone, anacordone, carbamidal, coramine, corvotone, nicamide
PubChem CID 5497
IUPAC Name N,N-diethylpyridine-3-carboxamide
SMILES CCN(CC)C(=O)C1=CN=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0600g/mL
Boiling Point 296°C to 300°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5240 to 1.526
Quantity 25 g
Beilstein 22, II, 34
Specific Gravity 1.06
Merck Index 15, 6627
Solubility Information Solubility in water: miscible. Other solubilities: miscible with methanol,ethanol,acetone,chlorofo,rm,ether
Formula Weight 178.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N, N-Diethylnicotinamide, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic in contact with skin.
Toxic if swallowed.
Toxic if inhaled.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-418-5
RTECSNumber QS4375000
Recommended Storage Product may darken on storage

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.