Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Dihydroxypyromellitimide 96.0+%, TCI America™
SDP

Catalog No. D44131G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44131G 1 g
D44135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D44131G Supplier TCI America Supplier No. D44131G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 57583-53-6
Molecular Formula C10H4N2O6
Molecular Weight (g/mol) 248.15
InChI Key VBBUVIWLZBAEEF-UHFFFAOYSA-N
PubChem CID 221479
IUPAC Name 2,6-dihydroxypyrrolo[3,4-f]isoindole-1,3,5,7-tetrone
SMILES C1=C2C(=CC3=C1C(=O)N(C3=O)O)C(=O)N(C2=O)O

Specifications

Color Yellow
Quantity 1 g
Synonym N,N′C-Dihydroxypyromellitic Diimide, NDHPI
Formula Weight 248.15
Percent Purity ≥96.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Dihydroxypyromellitimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.