Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethyl-3-nitroaniline 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
619-31-8
C8H10N2O2
166.18
MFCD00007236
CJDICMLSLYHRPT-UHFFFAOYSA-N
3-nitro-n,n-dimethylaniline,benzenamine, n,n-dimethyl-3-nitro,m-nitro-n,n-dimethylaniline,n,n-dimethyl-m-nitroaniline,1-dimethylamino-3-nitrobenzene,benzene, 1-dimethylamino-3-nitro,aniline, n,n-dimethyl-m-nitro,dimethyl-3-nitro-phenyl-amine,dimethyl 3-nitrophenyl amine,m-nitrodimethylaniline
69269
N,N-dimethyl-3-nitroaniline
CN(C)C1=CC(=CC=C1)[N+](=O)[O-]
619-31-8
C8H10N2O2
166.18
MFCD00007236
CJDICMLSLYHRPT-UHFFFAOYSA-N
3-nitro-n,n-dimethylaniline,benzenamine, n,n-dimethyl-3-nitro,m-nitro-n,n-dimethylaniline,n,n-dimethyl-m-nitroaniline,1-dimethylamino-3-nitrobenzene,benzene, 1-dimethylamino-3-nitro,aniline, n,n-dimethyl-m-nitro,dimethyl-3-nitro-phenyl-amine,dimethyl 3-nitrophenyl amine,m-nitrodimethylaniline
69269
N,N-dimethyl-3-nitroaniline
CN(C)C1=CC(=CC=C1)[N+](=O)[O-]
Color Orange to Orange-Red
Melting Point 57°C to 61°C
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula O2NC6H4N(CH3)2
Beilstein 12, 701
Solubility Information Solubility in water: insoluble
Formula Weight 166.18
Physical Form Crystals or Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material N, N-Dimethyl-3-nitroaniline, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 210-590-3

TSCA : TSCA