Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethyl-o-toluidine 99.0+%, TCI America™
SDP

Catalog No. d080525ml
Click to view available options
Quantity:
25 mL

Chemical Identifiers

CAS 609-72-3
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00035789
InChI Key JDEJGVSZUIJWBM-UHFFFAOYSA-N
Synonym n,n-dimethyl-o-toluidine, dimethyl-o-toluidine, o-methyldimethylaniline, benzenamine, n,n,2-trimethyl, o-toluidine, n,n-dimethyl, 2-methyl-n,n-dimethylaniline, n,n-dimethyl-2-methylaniline, benzene, 1-dimethylamino-2-methyl, n,n,2-trimethylbenzamine, n,n,2-trimethylbenzenamine
PubChem CID 11869
IUPAC Name N,N,2-trimethylaniline
SMILES CN(C)C1=CC=CC=C1C

Specifications

Color Yellow
Boiling Point 186°C
Quantity 25 mL
UN Number 2810
Formula Weight 135.21
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Dimethyl-o-toluidine
EINECSNumber (3)-0191
RTECSNumber XU5800000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.