Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethyl-p-nitroaniline, 99%, Thermo Scientific™

Catalog No. AC408410250
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-23-2
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00014712
InChI Key QJAIOCKFIORVFU-UHFFFAOYSA-N
Synonym 4-nitro-n,n-dimethylaniline, benzenamine, n,n-dimethyl-4-nitro, 4-nitrodimethylaniline, p-nitro-n,n-dimethylaniline, p-nitrodimethylaniline, 1-dimethylamino-4-nitrobenzene, 4-dimethylamino nitrobenzene, p-dimethylamino nitrobenzene, n,n-dimethyl-p-nitroaniline, aniline, n,n-dimethyl-p-nitro
PubChem CID 7491
IUPAC Name N,N-dimethyl-4-nitroaniline
SMILES CN(C)C1=CC=C(C=C1)[N+](=O)[O-]

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 162°C to 166°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3)2NC6H4NO2
Quantity 25 g
Formula Weight 166.18
Percent Purity ≥98.5%
Physical Form Amorphous Powder
Chemical Name or Material N, N-Dimethyl-p-nitroaniline
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Toxic if inhaled.
Toxic if swallowed.
Causes serious eye irritation.
Toxic in contact with skin.
Causes skin irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-832-1
RTECSNumber BX7035000
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.