Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N,N-Dimethyl-p-toluidine, 99%

Manufacturer:  Alfa Aesar™ A1198836

Quantity 500g

 View more versions of this product

Catalog No. AAA1198836


  Check Availability
Add to cart

Specifications

Description

Applications
N,N-Dimethyl-p-toluidine is used as a polymerization catalyst for polyesters, acrylate and epoxy resins. It is also used as a hardener for dental cements and in adhesives. It serves as an intermediate for photographic chemicals, in industrial glues, in artificial fingernail preparations, colorants, pharmaceuticals. It reacts with vinyl ether in the presence of copper(II) chloride gives tetrahydroquinolines. Further, it is used to accelerate polymerization of ethyl methacrylate.

Solubility
Miscible with alcohol, ether and chloroform. Immiscible with water.

Notes
Incompatible with strong oxidizing agents. Store in a cool place.
Description & Specifications

Chemical Identifiers

99-97-8
135.21
UN1708
N,N-Dimethyl-4-methylaniline; N,N,4-Trimethylaniline
C9H13N
MFCD00008316
774409
N,N-Dimethyl-p-toluidine

Specifications

99-97-8
210° to 211°C
83°C (181°F)
135.21
UN1708
N,N-Dimethyl-4-methylaniline; N,N,4-Trimethylaniline
99%
−25°C
500g
C9H13N
MFCD00008316
774409
N,N-Dimethyl-p-toluidine

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H311-H331-H373-H227
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.

GHS P Statement
P210-P260-P301+P310-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOLUIDINES, LIQUID

RTECSNumber : XU5803000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit