Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylacetamide, 99%

Catalog No. p-7081103
Click to view available options
Quantity:
250 mL
500 mL
2500 mL

CAS: 127-19-5 | C4H9NO | 87.12 g/mol

N,N-Dimethylacetamide is used as a polar solvent in organic synthesis as well as in reactions involving strong bases such as sodium hydroxide. It is also used as a solvent for fibers like polyacrylonitrile, spandex, as an excipient in drugs viz. vumon, busulfex and in the adhesive industry. It finds application as a reaction medium in the production of pharmaceuticals, agrochemicals, dyes and plasticizers. It is widely used in polymer industry due to its high solving power for high molecular-weight polymers and synthetic resins. It plays an important role as a catalyst in various reactions viz. cyclization, halogenation, cynidation, alkylation and dehydrogenation and increases the yield of main products. It is also used as an extracting agent for oil and gases, in paint removers, in the production of photo-resist stripping compounds and as booster solvent in coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylacetamide is used as a polar solvent in organic synthesis as well as in reactions involving strong bases such as sodium hydroxide. It is also used as a solvent for fibers like polyacrylonitrile, spandex, as an excipient in drugs viz. vumon, busulfex and in the adhesive industry. It finds application as a reaction medium in the production of pharmaceuticals, agrochemicals, dyes and plasticizers. It is widely used in polymer industry due to its high solving power for high molecular-weight polymers and synthetic resins. It plays an important role as a catalyst in various reactions viz. cyclization, halogenation, cynidation, alkylation and dehydrogenation and increases the yield of main products. It is also used as an extracting agent for oil and gases, in paint removers, in the production of photo-resist stripping compounds and as booster solvent in coatings.

Solubility
Miscible with water, benzene, alcohol, acetone, ether, esters and ketones.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-19-5
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00008686
InChI Key FXHOOIRPVKKKFG-UHFFFAOYSA-N
Synonym dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
PubChem CID 31374
ChEBI CHEBI:84254
IUPAC Name N,N-dimethylacetamide
SMILES CN(C)C(C)=O

Specifications

Melting Point -20°C
Density 0.94
Boiling Point 165°C to 166°C
Flash Point 70°C (158°F)
Odor Ammonia-like
Refractive Index 1.4385
Linear Formula CH3CON(CH3)2
Quantity 250 mL
Beilstein 1737614
Merck Index 14,3227
Solubility Information Miscible with water,benzene,alcohol,acetone,ether,esters and ketones.
Formula Weight 87.12
Percent Purity 99%
Chemical Name or Material N,N-Dimethylacetamide
Show More Show Less
Hazard Category H227-H312+H332-H360D
Hazard Statement GHS H Statement
H360-H312-H332-H227
May damage fertility or the unborn child.
Harmful in contact with skin.
Harmful if inhaled.
Combustible liquid.
Precautionary Statement P201-P202-P210-P261-P271-P281-P302+P352-P304+P340-P308+P313-P312-P363-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 204-826-4
RTECSNumber AB7700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.