Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylacetamide, 99.8+%, for HPLC, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
Packaging:
Glass bottle
127-19-5
C4H9NO
87.122
MFCD00008686
FXHOOIRPVKKKFG-UHFFFAOYSA-N
dimethylacetamide,dmac,acetamide, n,n-dimethyl,acetdimethylamide,dimethyl acetamide,n,n-dimethyl acetamide,dimethylamide acetate,n,n-dimethylethanamide,dimethylacetone amide,acetyldimethylamine
31374
CHEBI:84254
N,N-dimethylacetamide
CC(=O)N(C)C
127-19-5
C4H9NO
87.122
MFCD00008686
FXHOOIRPVKKKFG-UHFFFAOYSA-N
dimethylacetamide,dmac,acetamide, n,n-dimethyl,acetdimethylamide,dimethyl acetamide,n,n-dimethyl acetamide,dimethylamide acetate,n,n-dimethylethanamide,dimethylacetone amide,acetyldimethylamine
31374
CHEBI:84254
N,N-dimethylacetamide
CC(=O)N(C)C
For HPLC
Absorbance 0.046 at 310nm,0.301 at 285nm,0.7 (1 cm cell vs HPLC−grade water) at 275nm
Acidity 0.0005 meq/g max.
Alkalinity 0.003 meq/g max.
Assay Percent Range 99.8+%
Linear Formula CH3CON(CH3)2
Merck Index 15, 3248
Formula Weight 87.12
Physical Form Liquid
Percent Purity ≥99.8%
Grade HPLC
Density 0.9370g/mL
Infrared Spectrum Authentic
Non_volatile Matter 0.0003% max.
Packaging Glass bottle
Refractive Index 1.4370 to 1.4390
Solubility Solubility in water: soluble. Other solubilities: soluble in acetone, diethyl ether, benzene,, chloroform, ethanol and toluene
Specific Gravity 0.937
Water 0.05% max.
Flash Point 70°C
Viscosity 1.02 mPa.s (20°C)
Boiling Point 164.0°C to 166.0°C
Color Colorless
Melting Point -20.0°C
Quantity 1L
Chemical Name or Material N, N-Dimethylacetamide
Show More Show Less

GHS P Statement
May damage the unborn child.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger