Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylacetamide, 99%, Reagent Grade, Honeywell™

Catalog No. p-7120612 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-19-5
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00008686
InChI Key FXHOOIRPVKKKFG-UHFFFAOYSA-N
Synonym dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
PubChem CID 31374
ChEBI CHEBI:84254
IUPAC Name N,N-dimethylacetamide
SMILES CN(C)C(C)=O

Specifications

pH 4 (at 200.00g/L 20°C)
Melting Point -20°C
Color Colorless to Yellow
Density 0.937g/mL at 25°C
Boiling Point 164°C to 166°C (1013 hPa)
Quantity 2.5 L
Explosion Limit 11.5% (160°F), 1.8% (100°F)
Flash Point 70°C (158°F)
Ignition Point 914°F
Odor Amine-like
Packaging Glass Bottle
Physical Form Liquid
Linear Formula CH3CON(CH3)2
Vapor Pressure 4mmHg (38°C), 2mmHg (25°C)
Beilstein 1737614
Solubility Information Fully miscible in water
Formula Weight 87.12g/mol
Percent Purity 0.99
Chemical Name or Material N,N-Dimethylacetamide
Show More Show Less
Hazard Statement P201-P261-P280-P305 + P351 + P338-P308 + P313
RTECSNumber 204-826-4
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.