Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylacetamide (Certified), Fisher Chemical

Catalog No. d135500 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle

Chemical Identifiers

CAS 127-19-5
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00008686
InChI Key FXHOOIRPVKKKFG-UHFFFAOYSA-N
Synonym dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
PubChem CID 31374
ChEBI CHEBI:84254
IUPAC Name N,N-dimethylacetamide
SMILES CN(C)C(C)=O

Specifications

pH 4
Melting Point -20°C
Color Colorless
Density 0.937g/cm³
Boiling Point 164°C
Quantity 500 mL
Flash Point 87°C (188.6°F)
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Linear Formula CH3CON(CH3)2
Vapor Pressure 1.7mbar at 25°C
Viscosity 1.02 mPa/s at 20°C
UN Number 2810
Water 0.05% max.
Formula Weight 87.12
Grade Certified
Chemical Name or Material N, N-Dimethylacetamide
Show More Show Less
Hazard Category Signal Word: WARNING!
Precautionary Statement Emergency Overview
Combustible liquid. Harmful in contact with skin. Harmful by inhalation. Irritating to eyes. May cause harm to the unborn child.
NFPA
Health:2 Flammability:2 Instability:0
DOTInformation Class 6.1: Poison
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.