Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylacetamide dimethyl acetal, tech. 90%, stab.

Catalog No. p-7043622
Click to view available options
Quantity:
5 g
25 g

CAS: 18871-66-4 | C6H15NO2 | 133.191 g/mol

N,N-Dimethylacetamide dimethyl acetal is used as a reagent for the synthesis of amides, diacylamines, and heterocycles, one carbon inserting synthon in preparation of pyrimido[1,2-a][1,3,5]triazin-6-ones. It is used as intermediate in building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylacetamide dimethyl acetal is used as a reagent for the synthesis of amides, diacylamines, and heterocycles, one carbon inserting synthon in preparation of pyrimido[1,2-a][1,3,5]triazin-6-ones. It is used as intermediate in building block.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Store at 2 - 8°C. Incompatible with strong oxidizing agents, Acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18871-66-4
Molecular Formula C6H15NO2
Molecular Weight (g/mol) 133.191
MDL Number MFCD00008476
InChI Key FBZVZUSVGKOWHG-UHFFFAOYSA-N
Synonym n,n-dimethylacetamide dimethyl acetal, 1,1-dimethoxy-n,n-dimethylethylamine, 1,1-dimethoxyethyl dimethylamine, 1,1-dimethoxyethyl dimethyl amine, 1,1-dimethoxy-ethyl-dimethyl-amine, ethanamine, 1,1-dimethoxy-n,n-dimethyl, dimethylacetamide dimethylacetal, dimethylacetamide dimethyl acetal, n,n-dimethylacetamide dimethyl acetal, stabilized with methanol, zlchem 332
PubChem CID 87835
IUPAC Name 1,1-dimethoxy-N,N-dimethylethanamine
SMILES CC(N(C)C)(OC)OC

Specifications

Density 0.911
Boiling Point 118°C to 120°C
Flash Point 8°C (46°F)
Refractive Index 1.412
Quantity 5 g
UN Number UN1993
Beilstein 1420865
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 133.19
Percent Purity 90%
Grade Technical
Chemical Name or Material N,N-Dimethylacetamide dimethyl acetal, stabilized with 5 to 10% methanol
Show More Show Less
Hazard Category H225-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H224-H301-H311-H371-H332
Extremely flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 242-641-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.