Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium bisulfate monohydrate, 97%, extra pure, reagent, crystals

Catalog No. AC419430100
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic drum

CAS: 10034-88-5 | H3NaO5S | 138.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-88-5
Molecular Formula H3NaO5S
Molecular Weight (g/mol) 138.07
MDL Number MFCD00149210
InChI Key JXHZRQHZVYDRGX-UHFFFAOYSA-M
Synonym sodium bisulfate monohydrate, sodium hydrogen sulfate monohydrate, unii-3kls9y77yj, sulfuric acid, monosodium salt, hydrate, 3kls9y77yj, sodium hydrogensulfate hydrate, sulfuric acid, monosodium salt, monohydrate, sodium hydrate hydrogen sulfate, sodium hydrogen sulfate-1-hydrate, acmc-20ajox
PubChem CID 23673662
IUPAC Name sodium;hydrogen sulfate;hydrate
SMILES O.[Na+].OS([O-])(=O)=O

Specifications

Melting Point 58.0°C
Color White
Physical Form Adhering Crystals
Quantity 10 kg
Assay Percent Range 97%
Linear Formula NaHSO4·H2O
Merck Index 15,8722
Solubility Information Solubility in water: 670g/L. Other solubilities: decomposes by alcohol into sodium sulfate and, h2so4
Formula Weight 138.08
Percent Purity 97%
Grade Extra Pure
Packaging Plastic drum
Chemical Name or Material Sodium bisulfate monohydrate, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Precautionary Statement GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
RTECSNumber VZ1870000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.