Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylaminomethylferrocene 97.0+%, TCI America™
SDP

Catalog No. D127125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D127125G 25 g
D12715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D127125G Supplier TCI America Supplier No. D127125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1271-86-9
Molecular Formula C13H17FeN
Molecular Weight (g/mol) 243.13
MDL Number MFCD00001433
InChI Key JJJSTEANRWLZBH-UHFFFAOYNA-N
Synonym Ferrocenylmethyldimethylamine
IUPAC Name (Dimethylaminomethyl)ferrocene
SMILES [Fe].c1cccc1.CN(C)Cc1cccc1

Specifications

Color Brown
Quantity 25 g
Formula Weight 243.13
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N-Dimethylaminomethylferrocene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.