Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylaniline, 99%

Catalog No. p-7081773
Click to view available options
Quantity:
10,000 mL
100 mL
2500 mL
500 mL

CAS: 121-69-7 | C8H11N | 121.18 g/mol

N,N-Dimethylaniline acts as a precursor to triarylmethane dyes such as crystal violet and malachite green. It is also used in the synthesis of vanillin, methyl violet, and Michler's ketone. Further, it is used as a hardener for plastic resins and an acid scavenger in the manufacture of semisynthetic penicillins and cephalosporins. In addition to this, it is used as a promoter in the curing of polyester and vinyl ester resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylaniline acts as a precursor to triarylmethane dyes such as crystal violet and malachite green. It is also used in the synthesis of vanillin, methyl violet, and Michler′s ketone. Further, it is used as a hardener for plastic resins and an acid scavenger in the manufacture of semisynthetic penicillins and cephalosporins. In addition to this, it is used as a promoter in the curing of polyester and vinyl ester resins.

Solubility
Miscible with water, ethanol, ether, acetone, chloroform, benzene and alkaline.

Notes
Incompatible with strong oxidizing agents, Strong acids, acid chlorides, acid anhydrides, chloroformates and halogens.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-69-7
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00008304
InChI Key JLTDJTHDQAWBAV-UHFFFAOYSA-N
Synonym dimethylaniline, dimethylphenylamine, n,n-dimethylbenzenamine, benzenamine, n,n-dimethyl, dimethylamino benzene, n,n-dimethylphenylamine, n,n-dimethylbenzeneamine, dimethylaminobenzene, dwumetyloanilina, n,n-dimethylamino benzene
PubChem CID 949
ChEBI CHEBI:16269
IUPAC Name N,N-dimethylaniline
SMILES CN(C)C1=CC=CC=C1

Specifications

Melting Point 2°C
Density 0.956
pH 7.4
Boiling Point 193°C
Flash Point 62°C (143°F)
Odor Characteristic
Assay Percent Range 99%
Refractive Index 1.558
Quantity 100 mL
UN Number UN2253
Beilstein 507140
Merck Index 14,3234
Solubility Information Miscible with water,ethanol,ether,acetone,chloroform,benzene and alkaline.
Formula Weight 121.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N,N-Dimethylaniline
Show More Show Less
Hazard Category H227-H301+H311+H331-H351
Hazard Statement GHS H Statement
H301-H311-H331-H351-H227
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Suspected of causing cancer.
Combustible liquid.
Precautionary Statement P201-P202-P210-P235-P261-P264b-P270-P271-P281-P301+P310-P302+P352-P304+P340-P308+P313-P311-P312-P330-P361-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: N,N-DIMETHYLANILINE
EINECSNumber 204-493-5
RTECSNumber BX4725000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.