Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylaniline, 99%, ACROS Organics™

Click to view available options
Quantity:
5mL
1L
2.5L
10L
Packaging:
Glass Bottle
Plastic drum
121-69-7
C8H11N
121.18
MFCD00008304
JLTDJTHDQAWBAV-UHFFFAOYSA-N
dimethylaniline,dimethylphenylamine,n,n-dimethylbenzenamine,benzenamine, n,n-dimethyl,dimethylamino benzene,n,n-dimethylphenylamine,n,n-dimethylbenzeneamine,dimethylaminobenzene,dwumetyloanilina,n,n-dimethylamino benzene
949
CHEBI:16269
N,N-dimethylaniline
CN(C)C1=CC=CC=C1
121-69-7
C8H11N
121.18
MFCD00008304
JLTDJTHDQAWBAV-UHFFFAOYSA-N
dimethylaniline,dimethylphenylamine,n,n-dimethylbenzenamine,benzenamine, n,n-dimethyl,dimethylamino benzene,n,n-dimethylphenylamine,n,n-dimethylbenzeneamine,dimethylaminobenzene,dwumetyloanilina,n,n-dimethylamino benzene
949
CHEBI:16269
N,N-dimethylaniline
CN(C)C1=CC=CC=C1
Specific Gravity 0.95
Melting Point 1.5°C to 2.5°C
Flash Point 63°C
Refractive Index 1.5570 to 1.5590
Packaging Glass Bottle
Boiling Point 193.0°C to 194.0°C
Quantity 5mL
Density 0.9500g/mL
Infrared Spectrum Authentic
Color Yellow
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula C6H5N(CH3)2
Fieser 01,274; 02,147
Merck Index 15, 3256
Solubility Information Solubility in water: 1g/L (20°C). Other solubilities: freely soluble in alcohol, ether, chloroform, acet, carbon tetrachloride
Formula Weight 121.18
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material N, N-Dimethylaniline
Show More Show Less

GHS H Statement:
Toxic to aquatic life with long lasting effects.
Toxic in contact with skin.
Suspected of causing cancer.
Toxic if inhaled.
Toxic if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger