Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylbenzylamine, 99%

Catalog No. AC159750051
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
5 L
Packaging:
Glass bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC159750051 5 L Plastic drum
AC159751000 100 mL Glass bottle
AC159755000 500 mL Glass bottle
AC159750025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC159750051 Supplier Thermo Scientific Chemicals Supplier No. 159750051
Only null left
Add to Cart

CAS: 103-83-3 | C9H13N | 135.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-83-3
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00008329
InChI Key XXBDWLFCJWSEKW-UHFFFAOYSA-N
Synonym n,n-dimethylbenzylamine, benzyldimethylamine, n-benzyldimethylamine, dimethylbenzylamine, bdma, benzenemethanamine, n,n-dimethyl, benzyl-n,n-dimethylamine, n-phenylmethyl dimethylamine, n,n-dimethylbenzenemethanamine, araldite accelerator 062
PubChem CID 7681
SMILES CN(C)CC1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -75.0°C
Color Colorless to Yellow
Density 0.9000g/mL
Boiling Point 183.0°C to 184.0°C
Flash Point 54°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic drum
Refractive Index 1.5000 to 1.5020
Linear Formula C6H5CH2N(CH3)2
Quantity 5 L
Beilstein 12, 1019
Specific Gravity 0.9
Solubility Information Solubility in water: 8g/L (20°C). Other solubilities: soluble in alcohol,acetone,ether and toluene,reacts with organic and inorganic acids
IUPAC Name benzyldimethylamine
Formula Weight 135.23
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N, N-Dimethylbenzylamine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 203-149-1
RTECSNumber DP4500000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.