Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N,N-Dimethylethylenediamine

Catalog No. 39030500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
100 mL
500 mL

Catalog No. 39-030-500ML

Supplier: Sigma Aldrich 39030500ML

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • N,n-dimethylethylenediamine is a bidentate ligand that can be used:
  • To synthesize schiff base ligands to prepare their zinc alkyl and zinc alkoxide complexes that initiate ring-opening polymerization of lactides
  • As a ligand to prepare copper azido polymers
  • In the synthesis of naphthalimides such as amonafide, an antitumor agent
  • As the receptor in building fluorescent signaling systems to perturb the photoinduced intramolecular electron transfer (pet) process from the nitrogen lone-pair to the fluorophore

Specifications

CAS 108-00-9
Density 0.807 g/mL (at 20°C (literature))
Boiling Point 104°C to 106°C (lit.)
Molecular Formula C4H12N2
Refractive Index n20/D 1.426 (literature); n20/D 1.427
Linear Formula (CH3)2 NCH2CH2NH2
MDL Number MFCD00008175
Quantity 500 mL
Synonym 2-(Dimethylamino)ethylamine; DMEN
Molecular Weight (g/mol) 88.15
Percent Purity ≥98.0% (GC)
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.