Learn More
N,N-Dimethylethylamine, 99%
Description
N,N-Dimethylethylamine is used as adhesive, sealant chemical and process regulator. It acts as an intermediate for quaternary ammonium compounds and is also used as a stabilizer for chlorinated hydrocarbons and vinyl derivatives. It is used in foundries as a catalyst for sand core production. Further, it is employed in the polymerization of polyamides. In addition to this, it is used in the preparation of polypeptide nanogels with self-reinforced endocytoses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N,N-Dimethylethylamine is used as adhesive, sealant chemical and process regulator. It acts as an intermediate for quaternary ammonium compounds and is also used as a stabilizer for chlorinated hydrocarbons and vinyl derivatives. It is used in foundries as a catalyst for sand core production. Further, it is employed in the polymerization of polyamides. In addition to this, it is used in the preparation of polypeptide nanogels with self-reinforced endocytoses.
Solubility
Miscible with water.
Notes
Incompatible with oxidizing agents and peroxide.
Specifications
Specifications
| Melting Point | -140°C |
| Density | 0.675 |
| Boiling Point | 36°C to 37°C |
| Flash Point | −36°C (−33°F) |
| Odor | Ammonia-like |
| Refractive Index | 1.373 |
| Linear Formula | CH3CH2N(CH3)2 |
| Quantity | 100 mL |
| UN Number | UN2733 |
| Beilstein | 1696893 |
| Show More |
Safety and Handling
| Hazard Category | H225-H302+H332-H314 |
| Hazard Statement | GHS H Statement H225-H314-H318-H302-H332 Highly flammable liquid and vapour. Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. Harmful if inhaled. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S. |
| EINECSNumber | 209-940-8 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.