Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylethylenediamine 98.0+%, TCI America™
SDP

Catalog No. D0719100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0719100ML 100 mL
D071925ML 25 mL
D0719500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0719100ML Supplier TCI America Supplier No. D0719100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 108-00-9
Molecular Formula C4H12N2
Molecular Weight (g/mol) 88.154
MDL Number MFCD00008175
InChI Key DILRJUIACXKSQE-UHFFFAOYSA-N
Synonym n,n-dimethylethylenediamine, 2-dimethylaminoethylamine, 2-aminoethyl dimethylamine, 1,2-ethanediamine, n,n-dimethyl, 2-dimethylamino ethylamine, n,n-dimethyl-1,2-ethanediamine, n1,n1-dimethylethane-1,2-diamine, n,n-dimethyl-1,2-ethylenediamine, 2-aminoethyldimethylamine, n,n-dimethylethanediamine
PubChem CID 66053
IUPAC Name N',N'-dimethylethane-1,2-diamine
SMILES CN(C)CCN

Specifications

Color Yellow
Boiling Point 107°C
Quantity 100 mL
UN Number 2733
Formula Weight 88.15
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N-Dimethylethylenediamine
EINECSNumber (2)-0158
RTECSNumber KH8608250
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.