Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyl N-cyanocarbonimidate, 97%

Catalog No. AC297210100
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC297210100 10 g Glass bottle
AC297210500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC297210100 Supplier Thermo Scientific Chemicals Supplier No. 297210100
Only null left
Add to Cart

CAS: 79463-77-7 | C14H10N2O2 | 238.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79463-77-7
Molecular Formula C14H10N2O2
Molecular Weight (g/mol) 238.25
MDL Number MFCD00010380
InChI Key SLIKWVTWIGHFJE-UHFFFAOYSA-N
Synonym diphenyl n-cyanocarbonimidate, diphenyl cyanocarbonimidate, diphenoxymethylenecyanamide, n-cyano-diphenyl imidocarbonate, diphenylcyanocarbonimidate, cyanocarbonimidia acid diphenyl aester, n-cyanocarbonimidic acid diphenyl ester, bis phenoxy methylidenecyanamide, cyano diphenoxymethylidene amine, 3,3-diphenoxy-2-azaprop-2-enenitrile
PubChem CID 688090
SMILES N#CN=C(OC1=CC=CC=C1)OC1=CC=CC=C1

Specifications

Melting Point 155.0°C to 160.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min
Packaging Glass bottle
Linear Formula NCN=C(OC6H5)2
Quantity 10 g
Solubility Information Solubility in water: may decompose.
IUPAC Name cyano(diphenoxymethylidene)amine
Formula Weight 238.25
Percent Purity 97%
Physical Form Fluffy Powder
Chemical Name or Material Diphenyl N-cyanocarbonimidate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.