Learn More
N,N-Dimethylformamide, 99%
Description
N, N-Dimethylformamide is commonly used as a solvent. It is used as a reagent in Bouveault aldehyde synthesis and also in Vilsmeier-Haack reaction. It acts as a catalyst in the synthesis of acyl chlorides. It is used for separating and refining crude from olefin gas. DMF along with methylene chloride acts as a remover of varnish or lacquers. It is also used in the manufacture of adhesives, fibers and films.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N, N-Dimethylformamide is commonly used as a solvent. It is used as a reagent in Bouveault aldehyde synthesis and also in Vilsmeier-Haack reaction. It acts as a catalyst in the synthesis of acyl chlorides. It is used for separating and refining crude from olefin gas. DMF along with methylene chloride acts as a remover of varnish or lacquers. It is also used in the manufacture of adhesives, fibers and films.
Solubility
Miscible with water and most organic solvents.
Notes
Reaction with sodium hydride gives exothermic decomposition at low temperatures. Keep container tightly closed in a dry and well-ventilated place.

Specifications
Specifications
Melting Point | -61°C |
Density | 0.944 |
pH | 7 |
Boiling Point | 153°C |
Flash Point | 57°C (135°F) |
Odor | Amine-like |
Refractive Index | 1.431 |
Quantity | 250 mL |
UN Number | UN2265 |
Beilstein | 605365 |
Show More |
Safety and Handling
Hazard Category | H226-H312+H332-H319-H335-H336-H350-H360D |
Hazard Statement | GHS H Statement H360-H226-H312-H332-H319 May damage fertility or the unborn child. Flammable liquid and vapour. Harmful in contact with skin. Harmful if inhaled. Causes serious eye irritation. |
Precautionary Statement | P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P312-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: N,N-DIMETHYLFORMAMIDE |
EINECSNumber | 200-679-5 |
RTECSNumber | LQ2100000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.