Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylformamide, ≥99.8%, ACS Reagent, Honeywell Riedel-de Haën™

Catalog No. p-7120613 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle

ACS Reagent,≥99.8%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

Melting Point -61°C
Density 0.944g/mL
Boiling Point 153°C
Quantity 1 L
Explosion Limit 0.152
Flash Point 58°C (136.4°F)
Ignition Point 833°F
Packaging Glass Bottle
Linear Formula HCON(CH3)2
Vapor Pressure 2.7mmHg (20°C)
UN Number UN2265
Beilstein 605365
Formula Weight 73.09g/mol
Percent Purity ≥99.8%
Grade ACS Reagent
Chemical Name or Material N,N-Dimethylformamide
Show More Show Less
Hazard Statement P201-P210-P261-P280-P308 + P313-P370 + P378
Precautionary Statement H226-H312 + H332-H319-H360D
DOTInformation 3/PGIII
EINECSNumber 200-679-5
RTECSNumber LQ2100000
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.