Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylformamide-d{7}, 99.5% (Isotopic)

Catalog No. p-7051937
Click to view available options
Quantity:
1 g
5 g
10 g
5 x 1 g

CAS: 4472-41-7 | C3H7NO | 80.138 g/mol

It is a common solvent used in NMR spectroscopy. It is also used as a reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4472-41-7
Molecular Formula C3H7NO
Molecular Weight (g/mol) 80.138
MDL Number MFCD00003286
InChI Key ZMXDDKWLCZADIW-YYWVXINBSA-N
Synonym n,n-dimethylformamide-d7, dmf-d7, deuterated dmf, n,n-di 2h3 methyl 2h formamide, dimethyl formamide-d7, dimethylformamide d, heptadeutero-n,n-dimethylformamide, n,n-dimethylformamide-d7 dmf-d7, formamide-1-d, n,n-di methyl-d3, n,n-dimethylformamide-d7 99.5atom%d
PubChem CID 78225
IUPAC Name 1-deuterio-N,N-bis(trideuteriomethyl)formamide
SMILES CN(C)C=O

Specifications

Physical Form Liquid
Color Colorless
Quantity 1 g
Packaging Prescored Ampule
Density 1.04 g/mL
Flash Point 58°C (136°F)
Odor Amine-like
Boiling Point 153°C
Linear Formula DCON(CD3)2
UN Number UN2265
Solubility Information Soluble in water.
Formula Weight 80.14
Percent Purity 99.5%
Chemical Name or Material N,N-Dimethylformamide-d7, Isotopic
Show More Show Less
Hazard Category H226-H312+H332-H319-H360D
Hazard Statement GHS H Statement
H360-H226-H312-H332-H319
May damage fertility or the unborn child.
Flammable liquid and vapor.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Precautionary Statement P201-P202-P210-P233-P240-P241-P242-P243-P261-P264b-P271-P280i-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P312-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: N,N-DIMETHYLFORMAMIDE
EINECSNumber 224-745-8
TSCA No
Recommended Storage Ambient

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.