Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylformamide Dibutyl Acetal 98.0+%, TCI America™
SDP

Catalog No. D130225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D130225ML 25 mL
D13025ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D130225ML Supplier TCI America Supplier No. D130225ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18503-90-7
Molecular Formula C11H25NO2
Molecular Weight (g/mol) 203.33
MDL Number MFCD00015249
InChI Key GSFFXKGTGPMVLM-UHFFFAOYSA-N
Synonym 1,1-Dibutoxytrimethylamine
PubChem CID 87683
IUPAC Name (dibutoxymethyl)dimethylamine
SMILES CCCCOC(OCCCC)N(C)C

Specifications

Color Yellow
Boiling Point 98°C
Quantity 25 mL
UN Number 1993
Formula Weight 203.33
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N-Dimethylformamide Dibutyl Acetal [for Esterification]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.