Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylformamide dimethyl acetal, 94%

Catalog No. p-7022527
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 4637-24-5 | C5H13NO2 | 119.16 g/mol

N,N-Dimethylformamide dimethyl acetal is used as an intermediate in the formation of pyridine derivatives, which exhibits inhibition against PI3 kinase p110alfa enzymes. It is utilized for the derivatization of primary sulfonamides and trifluoroacetic acid. It is also used in the preparation of formamidine derivatives. It is used as a reagent for n-dimethylaminomethylene and methyl esters. Further, it is used to catalyze the coupling of epoxides with carbon dioxide to prepare cyclic carbonates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylformamide dimethyl acetal is used as an intermediate in the formation of pyridine derivatives, which exhibits inhibition against PI3 kinase p110alfa enzymes. It is utilized for the derivatization of primary sulfonamides and trifluoroacetic acid. It is also used in the preparation of formamidine derivatives. It is used as a reagent for n-dimethylaminomethylene and methyl esters. Further, it is used to catalyze the coupling of epoxides with carbon dioxide to prepare cyclic carbonates.

Solubility
Miscible with most organic solvents.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4637-24-5
Molecular Formula C5H13NO2
Molecular Weight (g/mol) 119.16
MDL Number MFCD00008482
InChI Key ZSXGLVDWWRXATF-UHFFFAOYSA-N
Synonym n,n-dimethylformamide dimethyl acetal, 1,1-dimethoxytrimethylamine, dimethoxymethyl dimethylamine, methanamine, 1,1-dimethoxy-n,n-dimethyl, dimethylformamide-dimethylacetal, dmf dimethyl acetal, dmfdma, dimethylformamide dimethyl acetal, 1,1-dimethoxy-n,n-dimethylmethylamine, dimethoxy dimethylamino methane
PubChem CID 78373
ChEBI CHEBI:85061
IUPAC Name (dimethoxymethyl)dimethylamine
SMILES COC(OC)N(C)C

Specifications

Melting Point -85°C
Density 0.897
Boiling Point 102°C to 104°C
Flash Point 7°C (45°F)
Refractive Index 1.3972
Quantity 25 g
UN Number UN1993
Sensitivity Moisture sensitive
Solubility Information Miscible with most organic solvents.
Formula Weight 119.16
Percent Purity 97%
Chemical Name or Material N,N-Dimethylformamide dimethyl acetal
Show More Show Less
Hazard Category H225-H317-H318-H332-H361
Hazard Statement GHS H Statement
H225-H302-H332-H315-H319-H335
Highly flammable liquid and vapour.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P233-P240-P241-P242-P243-P261-P271-P272-P280g-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P312-P333+P313-P363-P370+P378q-P372-P374-P380-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 225-063-3
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.