Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylformamide dimethyl acetal, ca. 97%

Catalog No. p-117515
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC161250250 25 g Glass bottle
AC161251000 100 g Glass bottle
AC161255000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC161250250 Supplier Thermo Scientific Chemicals Supplier No. 161250250
Only null left
Add to Cart

CAS: 4637-24-5 | C5H13NO2 | 119.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4637-24-5 , 149-73-5 , 68-12-2 , 67-56-1
Molecular Formula C5H13NO2
Molecular Weight (g/mol) 119.16
MDL Number MFCD00008482
InChI Key ZSXGLVDWWRXATF-UHFFFAOYSA-N
Synonym n,n-dimethylformamide dimethyl acetal, 1,1-dimethoxytrimethylamine, dimethoxymethyl dimethylamine, methanamine, 1,1-dimethoxy-n,n-dimethyl, dimethylformamide-dimethylacetal, dmf dimethyl acetal, dmfdma, dimethylformamide dimethyl acetal, 1,1-dimethoxy-n,n-dimethylmethylamine, dimethoxy dimethylamino methane
PubChem CID 78373
ChEBI CHEBI:85061
IUPAC Name (dimethoxymethyl)dimethylamine
SMILES COC(OC)N(C)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 0.8900g/mL
Boiling Point 102.0°C to 104.0°C
Flash Point 7°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.3972
Linear Formula (CH3)2NCH(OCH3)2
Quantity 25 g
UN Number 1993
Fieser 03,115; 04,184; 05,254; 06,221; 08,191; 09,182; 10,158; 13,120; 05,646; 06,566; 07,355; 09,447; 11,501
Specific Gravity 0.89
Solubility Information Solubility in water: hydrolysis. Other solubilities: soluble in many organic solvents
Formula Weight 119.16
Percent Purity ca. 97%
Physical Form Liquid
Chemical Name or Material N, N-Dimethylformamide dimethyl acetal, ca 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye damage.
May cause an allergic skin reaction.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF exposed or concerned: Get medical attention/advice.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face p
EINECSNumber 225-063-3
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.