Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylformamide Dipropyl Acetal 90.0+%, TCI America™
SDP

Catalog No. D130125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D130125ML 25 mL
D13015ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D130125ML Supplier TCI America Supplier No. D130125ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6006-65-1
Molecular Formula C9H22NO2
Molecular Weight (g/mol) 176.28
MDL Number MFCD00009374
InChI Key NSLGQFIDCADTAS-UHFFFAOYSA-O
Synonym n,n-dimethylformamide dipropyl acetal, 1,1-dipropoxytrimethylamine, n,n-dimethylformamide di-n-propyl acetal, dipropoxymethyl dimethylamine, 1,1-dipropoxy-n,n-dimethylmethylamine, acmc-1b5t3, dimethyl dipropoxymethyl amine, dimethylformamide dipropyl acetal, nslgqfidcadtas-uhfffaoysa, n,n-dimethylformamide dipropylacetal
PubChem CID 80105
IUPAC Name (dipropoxymethyl)dimethylazanium
SMILES CCCOC(OCCC)[NH+](C)C

Specifications

Color Yellow
Quantity 25 mL
UN Number 1993
Formula Weight 175.27
Percent Purity ≥90.0% (T)
Physical Form Liquid
Chemical Name or Material N,N-Dimethylformamide Dipropyl Acetal [for Esterification]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.