Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylformamide, For gas chromatography ECD and FID SupraSolv™, MilliporeSigma™

Catalog No. p-7020449 Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle

For gas chromatography ECD and FID SupraSolv™

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

pH 7 (at 200g/L 20°C)
Melting Point -61°C
Color Colorless
Density 0.94g/cm3 at 20°C
Boiling Point 153°C
Quantity 1 L
Flash Point 58°C (136°F)
Ignition Point 410°C (770°F)
Odor Weakly amine-like
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.43
Vapor Pressure 3.77 hPa (20°C)
UN Number 2265
Solubility Information Soluble in water at 20°C
Formula Weight 73.09
Chemical Name or Material N,N-Dimethylformamide
Show More Show Less
Hazard Category H360D May damage the unborn child.
Hazard Statement H226 Flammable liquid and vapor.
Precautionary Statement H312 + H332 Harmful in contact with skin or if inhaled.
Recommended Storage 5°C to 30°C (41°F to 86°F); Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition. Keep locked up or in an area accessible only to qualified or authorized persons.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.