Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N,N-Dimethylthioformamide, 97%
Click to view available options
Quantity:
5 g
25 g
100 g
Description
It is used in telecommunication.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in telecommunication.
Solubility
Fully miscible in water.
Notes
Store at room temperature. Incompatible with oxidizing agents.
It is used in telecommunication.
Solubility
Fully miscible in water.
Notes
Store at room temperature. Incompatible with oxidizing agents.

Specifications
Specifications
Boiling Point | 58°C to 60°C (1 mmHg) |
Quantity | 25 g |
Solubility Information | Fully miscible in water. |
Formula Weight | 89.15 |
Percent Purity | 97% |
Chemical Name or Material | N,N-Dimethylthioformamide |
Safety and Handling
Hazard Category | H312+H332 |
Hazard Statement | GHS H Statement H301-H311-H332-H315-H319-H335 Toxic if swallowed. Toxic in contact with skin. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P271-P280-P302+P352-P304+P340-P312-P363-P501c |
DOTInformation | Hazard Class: 6.1; Packaging Group: III |
EINECSNumber | 212-060-7 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction