Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Dinitrosopenta-methylenetetramine, Pure, MP Biomedicals

Catalog No. ICN206314
Click to view available options
Quantity:
10 g

Chemical Identifiers

CAS 101-25-7
Molecular Formula C5H10N6O2
Molecular Weight (g/mol) 186.175
InChI Key MWRWFPQBGSZWNV-UHFFFAOYSA-N
Synonym dipentax, micropor, dnpmt, opex, mikrofor n, porophor b, dinitrosopentamethylenetetramine, vulcacel bn, unicel-nd, unicel ndx
PubChem CID 7549
ChEBI CHEBI:82457
IUPAC Name 3,7-dinitroso-1,3,5,7-tetrazabicyclo[3.3.1]nonane
SMILES C1N2CN(CN1CN(C2)N=O)N=O

Specifications

Quantity 10 g
Formula Weight 186.2
Chemical Name or Material N, N«-Dinitrosopenta-Methylenetetramine, Pure
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.