Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N',N″,N‴-Tetraacetylglycoluril 98.0+%, TCI America™
SDP

Catalog No. T098525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T098525G 25 g
T09855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T098525G Supplier TCI America Supplier No. T098525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10543-60-9
Molecular Formula C12H14N4O6
Molecular Weight (g/mol) 310.27
MDL Number MFCD00022618
InChI Key KIHGYZTVBURVBA-UHFFFAOYSA-N
Synonym n,n',n,n'-tetraacetylglycoluril, 1,3,4,6-tetraacetyltetrahydroimidazo 4,5-d imidazole-2,5 1h,3h-dione, imidazo 4,5-d imidazole-2,5 1h,3h-dione, 1,3,4,6-tetraacetyltetrahydro, n,n',n,n'-tetraacetylacetyleneurea, n,n',n,n'-tetraacetylglycouril, 1,3,4,6-tetraacetyl-dihydro-1,3 diazolo 4,5-d imidazole-2,5-dione, tetraacetylglycoluril, tetraacetylglykoluril
PubChem CID 82717
IUPAC Name 1,3,4,6-tetraacetyl-octahydro-[1,3]diazolo[4,5-d]imidazole-2,5-dione
SMILES CC(=O)N1C2C(N(C(C)=O)C1=O)N(C(C)=O)C(=O)N2C(C)=O

Specifications

Melting Point 237°C
Color White
Quantity 25 g
Formula Weight 310.27
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′,N′′,N′′′-Tetraacetylglycoluril
EINECSNumber (5)-1115
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.