Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-N,N,N',N'-Tetramethyl-L-tartardiamide 98.0+%, TCI America™
SDP

Catalog No. T183925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T183925G 25 g
T18395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T183925G Supplier TCI America Supplier No. T183925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 26549-65-5
Molecular Formula C8H16N2O4
Molecular Weight (g/mol) 204.226
MDL Number MFCD00025672
InChI Key PCYDYHRBODKVEL-PHDIDXHHSA-N
PubChem CID 6610750
IUPAC Name (2R,3R)-2,3-dihydroxy-N,N,N',N'-tetramethylbutanediamide
SMILES CN(C)C(=O)C(C(C(=O)N(C)C)O)O

Specifications

Melting Point 187°C
Color White
Quantity 25 g
Synonym (R,R)-(+)-2,3-Dihydroxy-N,N,N′,N′-tetramethylsuccinamide
Formula Weight 204.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (+)-N,N,N′,N′-Tetramethyl-L-tartardiamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.