Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclopentylmagnesium bromide, 2.0M solution in diethyl ether, AcroSeal™
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Gray |
Density | 0.9960g/mL |
Boiling Point | 35°C |
Flash Point | -38°C |
Packaging | AcroSeal™ Glass Bottle |
Quantity | 100 mL |
Specific Gravity | 0.996 |
Formula Weight | 173.33 |
Physical Form | Solution |
Chemical Name or Material | Cyclopentylmagnesium bromide, 2.0M solution in diethyl ether |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Harmful if swallowed. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. Extremely flammable liquid and vapor. In contact with water releases flammable gas. May form explosive peroxides. Reacts violently with water. Repeated exposure may cause skin dryness or cracking. |
Precautionary Statement | GHS P Statement Wear eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Avoid breathing dust/fume/gas/mist/vapors/spray. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Store in a dry place. Store in a closed container. |
EINECSNumber | 251-420-8 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction