Learn More
N,N,N',N'-Tetramethylethylenediamine, 99%
Description
N,N,N',N'-Tetramethylethylenediamine is used with ammonium persulfate to catalyze the polymerization of acrylamide and bis-acrylamide to get polyacrylamide gels. It is also used for the separation of proteins or nucleic acids. Further, it is employed as a ligand for metal ions like zinc and copper. It is actively involved in the formation of anionic organometallic complex.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N,N,N′,N′-Tetramethylethylenediamine is used with ammonium persulfate to catalyze the polymerization of acrylamide and bis-acrylamide to get polyacrylamide gels. It is also used for the separation of proteins or nucleic acids. Further, it is employed as a ligand for metal ions like zinc and copper. It is actively involved in the formation of anionic organometallic complex.
Solubility
Miscible with water.
Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, carbon dioxide and copper.
Specifications
Specifications
| Melting Point | °C to 55°C |
| Density | 0.775 |
| Boiling Point | 120°C to 122°C |
| Flash Point | 18°C (62°F) |
| Assay Percent Range | 99% |
| Refractive Index | 1.418 |
| Linear Formula | (CH3)2NCH2CH2N(CH3)2 |
| Quantity | 100 mL |
| UN Number | UN2372 |
| Beilstein | 1732991 |
| Show More |
Safety and Handling
| Hazard Category | H225-H302+H332-H314-H335 |
| Hazard Statement | GHS H Statement H225-H314-H302-H332 |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 1,2-DI-(DIMETHYLAMINO)ETHANE |
| EINECSNumber | 203-744-6 |
| RTECSNumber | KV7175000 |
| TSCA | Yes |
| storageNote1 | Hygroscopic |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.