Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,N',N'-Tetramethylmethylenediamine, 99%

Catalog No. p-7044011
Click to view available options
Quantity:
25 g
100 g

CAS: 51-80-9 | C5H14N2 | 102.18 g/mol

N,N,N',N'-Tetramethylmethylenediamine is a reagent used as a convenient source of the Mannich intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N,N′,N′-Tetramethylmethylenediamine is a reagent used as a convenient source of the Mannich intermediate.

Solubility
Fully miscible in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-80-9
Molecular Formula C5H14N2
Molecular Weight (g/mol) 102.18
MDL Number MFCD00008328
InChI Key VGIVLIHKENZQHQ-UHFFFAOYSA-N
Synonym n,n,n',n'-tetramethyldiaminomethane, bis dimethylamino methane, methanediamine, n,n,n',n'-tetramethyl, tetramethylmethylenediamine, methylenebis dimethylamine, n,n,n',n'-tetramethylmethylenediamine, unii-z870i525ks, methylenediamine, n,n,n',n'-tetramethyl, dimethylamino methyl dimethylamine, dimethyl dimethylamino methyl amine
PubChem CID 5829
SMILES CN(C)CN(C)C

Specifications

Melting Point -55°C
Density 0.749
Boiling Point 83°C to 85°C
Flash Point −12°C (10°F)
Refractive Index 1.401
Linear Formula (CH3)2NCH2N(CH3)2
Quantity 25 g
UN Number UN2733
Beilstein 1731946
Solubility Information Fully miscible in water.
IUPAC Name N,N,N',N'-tetramethylmethanediamine
Formula Weight 102.18
Percent Purity 99%
Chemical Name or Material N,N,N',N'-Tetramethylmethylenediamine
Show More Show Less
Hazard Category H225-H314-H335
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 200-124-7
RTECSNumber PA6700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.