Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,N',N'-Tetramethylthiourea, 98%

Catalog No. aal1339206
Click to view available options
Quantity:
5 g
25 g

CAS: 2782-91-4 | C5H12N2S | 132.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2782-91-4
Molecular Formula C5H12N2S
Molecular Weight (g/mol) 132.23
MDL Number MFCD00008324
InChI Key MNOILHPDHOHILI-UHFFFAOYSA-N
Synonym tetramethylthiourea, thiourea, tetramethyl, basthioryl, 1,1,3,3-tetramethyl-2-thiourea, tmtu, n,n,n',n'-tetramethylthiourea, urea, thio-, tetramethyl, tetramethyl-2-thiourea, urea, 1,1,3,3-tetramethyl-2-thio, thiourea, n,n,n',n'-tetramethyl
PubChem CID 17725
IUPAC Name 1,1,3,3-tetramethylthiourea
SMILES CN(C)C(=S)N(C)C

Specifications

Melting Point 75°C to 77°C
Boiling Point 245°C
Flash Point 190°C (374°F)
Odor Odorless
Quantity 5 g
Beilstein 1744916
Formula Weight 132.23
Percent Purity 98%
Chemical Name or Material N,N,N',N'-Tetramethylthiourea
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 220-488-0
RTECSNumber YU2750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.