Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,N'-Triethylethylenediamine 98.0+%, TCI America™
SDP

Catalog No. T074325ML
Click to view available options
Quantity:
5 mL
25 mL

Chemical Identifiers

CAS 105-04-4
Molecular Formula C8H20N2
Molecular Weight (g/mol) 144.26
MDL Number MFCD00009054
InChI Key HDCAZTXEZQWTIJ-UHFFFAOYSA-N
Synonym n,n,n'-triethylethylenediamine, triethylethylenediamine, n,n,n'-triethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n'-triethyl, 2-diethylamino ethyl ethyl amine, 1,2-ethanediamine, n1,n1,n2-triethyl, ethylenediamine, n,n,n'-triethyl, n1,n1,n2-triethyl-1,2-ethanediamine, n,n,n-triethylenediamine, diethyl 2-ethylaminoethyl amine
PubChem CID 66040
IUPAC Name [2-(diethylamino)ethyl](ethyl)amine
SMILES CCNCCN(CC)CC

Specifications

Color Yellow
Boiling Point 145°C
Quantity 25 mL
UN Number 2734
Formula Weight 144.26
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N,N′-Triethylethylenediamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.