Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Nitrosodimethylamine Solution, 100μg/mL in Methanol, Ultra Scientific

Catalog No. usns1001
Encompass
Click to view available options
Quantity:
1 mL

Catalog No. US-NS-1001

Supplier: Agilent Technologies NS1001

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 62-75-9, 67-56-1
Molecular Formula C2H6N2O
Molecular Weight (g/mol) 74.083
InChI Key UMFJAHHVKNCGLG-UHFFFAOYSA-N
PubChem CID 6124
ChEBI CHEBI:35807
IUPAC Name N,N-dimethylnitrous amide
SMILES CN(C)N=O

Specifications

CAS Max % 99.99, 0.01
Physical Form Liquid
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 100μg/mL
Chemical Name or Material N-Nitrosodimethylamine Solution, 100μg/mL
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.