Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Nonanoic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
1kg
2.5kg
Packaging:
Glass bottle
112-05-0
C9H18O2
158.24
MFCD00004433
FBUKVWPVBMHYJY-UHFFFAOYSA-N
pelargonic acid,n-nonanoic acid,nonylic acid,pelargic acid,nonoic acid,n-nonylic acid,n-nonoic acid,1-octanecarboxylic acid,pelargon,cirrasol 185a
8158
CHEBI:29019
nonanoic acid
CCCCCCCCC(=O)O
112-05-0
C9H18O2
158.24
MFCD00004433
FBUKVWPVBMHYJY-UHFFFAOYSA-N
pelargonic acid,n-nonanoic acid,nonylic acid,pelargic acid,nonoic acid,n-nonylic acid,n-nonoic acid,1-octanecarboxylic acid,pelargon,cirrasol 185a
8158
CHEBI:29019
nonanoic acid
CCCCCCCCC(=O)O
Specific Gravity 0.9
Flash Point 114°C
Refractive Index 1.4299 to 1.4339
Infrared Spectrum Authentic
Packaging Glass bottle
Color Colorless
Quantity 100g
Melting Point 12.5°C
Boiling Point 254.0°C
Density 0.9000g/mL
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3(CH2)7CO2H
Beilstein 02, 352
Merck Index 15, 7179
Solubility Information Solubility in water: negligible. Other solubilities: soluble in alcohol, chloroform and ether, soluble in most other commmon organic solvents
Formula Weight 158.24
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material n-Nonanoic acid
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger