Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Nonylbenzene, 97%

Catalog No. aaa1295709
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 1081-77-2 | C15H24 | 204.357 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1081-77-2
Molecular Formula C15H24
Molecular Weight (g/mol) 204.357
MDL Number MFCD00009575
InChI Key LIXVMPBOGDCSRM-UHFFFAOYSA-N
Synonym 1-phenylnonane, benzene, nonyl, n-nonylbenzene, nonane, 1-phenyl, 1-nonylbenzene, unii-58ak68ov26, benzene, c6-12-alkyl derivs., n-nonyl benzene, phenyl-nonane, c6-c12 alkylbenzene
PubChem CID 14126
IUPAC Name nonylbenzene
SMILES CCCCCCCCCC1=CC=CC=C1

Specifications

Density 0.858
Boiling Point 281°C to 283°C
Flash Point >110°C (230°F)
Refractive Index 1.483
Quantity 10 g
Beilstein 1907173
Formula Weight 204.36
Percent Purity 97%
Chemical Name or Material n-Nonylbenzene
EINECSNumber 214-103-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.