Learn More
R&D Systems™ N-Nonyldeoxynojirimycin
GREENER_CHOICESDP
Catalog No. 369110R Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
Catalog No. Quantity
36-911-0R 10 mg
1 options
This item is not returnable. View return policy
Catalog No. 36-911-0R Supplier R&D Systems™ Supplier No. 3691/10
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
A Tocris product, part of R&D Systems. Glucosidase inhibitor
N-Nonyldeoxynojirimycin is a glucosidase inhibitor (IC50 values are 0.42 and 8.4 μM for acid α-glucosidase and α-1,6-glucosidase respectively). Inhibits liver glycogen breakdown in vivo. Also acts as a chemical chaperone; chaperones β-Glu folding at neutral p.H. allowing the stabilized enzyme to transit from the endoplasmic reticulum to the golgi, enabling proper trafficking to the lysosome.
Specifications
Quantity 10 mg
Product Line Tocris
Product Type N-Nonyldeoxynojirimycin
Molecular Weight (g/mol) 289.41
Inhibitors N-Nonyldeoxynojirimycin