Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,O-Bis(trimethylsilyl)acetamide 80.0+%, TCI America™
SDP

Catalog No. B0511100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0511100ML 100 mL
B051110ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0511100ML Supplier TCI America Supplier No. B0511100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10416-59-8
Molecular Formula C8H21NOSi2
Molecular Weight (g/mol) 203.43
MDL Number MFCD00008270
InChI Key SIOVKLKJSOKLIF-UHFFFAOYSA-N
Synonym n,o-bis trimethylsilyl acetamide, trimethylsilyl 1z-n-trimethylsilylethanimidate
PubChem CID 6913588
IUPAC Name trimethylsilyl N-(trimethylsilyl)ethanimidate
SMILES CC(O[Si](C)(C)C)=N[Si](C)(C)C

Specifications

Melting Point -24°C
Color Yellow
Boiling Point 73°C
Quantity 100 mL
UN Number 2920
Formula Weight 203.43
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material N,O-Bis(trimethylsilyl)acetamide
EINECSNumber (2)-3273
RTECSNumber AK3000000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.