Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,O-Bis(trimethylsilyl)hydroxylamine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
22737-37-7
C6H19NOSi2
177.4
MFCD00008266
ZAEUMMRLGAMWKE-UHFFFAOYSA-N
n,o-bis trimethylsilyl hydroxylamine,1,1,1-trimethyl-n-trimethylsilyl oxy silylamine,silanamine, 1,1,1-trimethyl-n-trimethylsilyl oxy,2,2,5,5-tetramethyl-3-oxa-4-aza-2,5-disilahexane,acmc-209fyl,hydroxylamine, 2tms derivative,n,o-bis-trimethylsilylhydroxylamine,n,o-bis trimethylsilyl hydroxylamin,n,o-bis-trimethylsilyl hydroxylamine
89815
[dimethyl-(trimethylsilyloxyamino)silyl]methane
C[Si](C)(C)NO[Si](C)(C)C
22737-37-7
C6H19NOSi2
177.4
MFCD00008266
ZAEUMMRLGAMWKE-UHFFFAOYSA-N
n,o-bis trimethylsilyl hydroxylamine,1,1,1-trimethyl-n-trimethylsilyl oxy silylamine,silanamine, 1,1,1-trimethyl-n-trimethylsilyl oxy,2,2,5,5-tetramethyl-3-oxa-4-aza-2,5-disilahexane,acmc-209fyl,hydroxylamine, 2tms derivative,n,o-bis-trimethylsilylhydroxylamine,n,o-bis trimethylsilyl hydroxylamin,n,o-bis-trimethylsilyl hydroxylamine
89815
[dimethyl-(trimethylsilyloxyamino)silyl]methane
C[Si](C)(C)NO[Si](C)(C)C
Specific Gravity 0.83
Quantity 1g
Boiling Point 78.0°C to 80.0°C (100.0mmHg)
Infrared Spectrum Authentic
Flash Point 28°C
Color Colorless
Packaging Glass bottle
Refractive Index 1.4112
Density 0.8300g/mL
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula (CH3)3SiONHSi(CH3)3
Fieser 06,58; 08,523; 09,512; 06,183; 10,142; 12,183; 13,110; 16,128
Formula Weight 177.4
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material N, O-Bis(trimethylsilyl)hydroxylamine, 97%
Show More Show Less

GHS H Statement
Flammable liquid and vapor.
Causes severe skin burns and eye damage.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin

GHS Signal Word: Danger

EINECSNumber : 245-188-7