Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,O-Bis(trimethylsilyl)hydroxylamine, 97%

Catalog No. AC215980050
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

CAS: 22737-37-7 | C6H19NOSi2 | 177.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22737-37-7
Molecular Formula C6H19NOSi2
Molecular Weight (g/mol) 177.39
MDL Number MFCD00008266
InChI Key ZAEUMMRLGAMWKE-UHFFFAOYSA-N
Synonym n,o-bis trimethylsilyl hydroxylamine, 1,1,1-trimethyl-n-trimethylsilyl oxy silylamine, silanamine, 1,1,1-trimethyl-n-trimethylsilyl oxy, 2,2,5,5-tetramethyl-3-oxa-4-aza-2,5-disilahexane, acmc-209fyl, hydroxylamine, 2tms derivative, n,o-bis-trimethylsilylhydroxylamine, n,o-bis trimethylsilyl hydroxylamin, n,o-bis-trimethylsilyl hydroxylamine
PubChem CID 89815
SMILES C[Si](C)(C)NO[Si](C)(C)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 0.8300g/mL
Boiling Point 78.0°C to 80.0°C (100.0 mmHg)
Flash Point 28°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4112
Linear Formula (CH3)3SiONHSi(CH3)3
Quantity 5 g
Fieser 06,58; 08,523; 09,512; 06,183; 10,142; 12,183; 13,110; 16,128
Specific Gravity 0.83
IUPAC Name [dimethyl-(trimethylsilyloxyamino)silyl]methane
Formula Weight 177.4
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N, O-Bis(trimethylsilyl)hydroxylamine, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
EINECSNumber 245-188-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.