Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,O-Dimethylhydroxylamine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. D1899100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1899100G 100 g
D189925G 25 g
D1899500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D1899100G Supplier TCI America Supplier No. D1899100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6638-79-5
Molecular Formula C2H8ClNO
Molecular Weight (g/mol) 97.54
MDL Number MFCD00012485
InChI Key USZLCYNVCCDPLQ-UHFFFAOYSA-N
Synonym n,o-dimethylhydroxylamine hydrochloride, n,o-dimethylhydroxylamine hcl, n-methoxymethylamine hydrochloride, methanamine, n-methoxy-, hydrochloride, o,n-dimethylhydroxylamine hydrochloride, n-methoxymethanamine hydrochloride, o,n-dimethyl-hydroxylamine hydrochloride, methoxy methyl amine hydrochloride, n,o-dimethylhydroxyamine hydrochloride, n,o-dimethylhydroxylaminehydrochloride
PubChem CID 81138
IUPAC Name hydrogen methoxy(methyl)amine chloride
SMILES [H+].[Cl-].CNOC

Specifications

Melting Point 115°C
Color White
Quantity 100 g
Formula Weight 97.54
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N,O-Dimethylhydroxylamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.