Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Octadecyl mercaptan, 96%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
2885-00-9
C18H38S
286.56
MFCD00004886
QJAOYSPHSNGHNC-UHFFFAOYSA-N
octadecanethiol,1-octadecanethiol,stearyl mercaptan,n-octadecyl mercaptan,octadecyl mercaptan,1-mercaptooctadecane,octadecanthiol,n-octadecanethiol,octadecanethol,stearylmercaptan
17905
octadecane-1-thiol
CCCCCCCCCCCCCCCCCCS
2885-00-9
C18H38S
286.56
MFCD00004886
QJAOYSPHSNGHNC-UHFFFAOYSA-N
octadecanethiol,1-octadecanethiol,stearyl mercaptan,n-octadecyl mercaptan,octadecyl mercaptan,1-mercaptooctadecane,octadecanthiol,n-octadecanethiol,octadecanethol,stearylmercaptan
17905
octadecane-1-thiol
CCCCCCCCCCCCCCCCCCS
Melting Point 25.0°C to 35.0°C
Flash Point 185°C
Boiling Point 366.0°C
Packaging Glass bottle
Quantity 25g
Refractive Index 1.4645
Infrared Spectrum Authentic
Color White
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Linear Formula CH3(CH2)16CH2SH
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in organic solvents
Formula Weight 286.56
Physical Form Crystalline Solid
Percent Purity 96%
Chemical Name or Material n-Octadecyl mercaptan
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning