Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Octanohydrazide 97.0+%, TCI America™
SDP

Catalog No. O032425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O032425G 25 g
O03245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O032425G Supplier TCI America Supplier No. O032425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6304-39-8
Molecular Formula C8H18N2O
Molecular Weight (g/mol) 158.25
MDL Number MFCD00011588
InChI Key VMUZVGRNTPFTKE-UHFFFAOYSA-N
Synonym n-Octanoic Hydrazide
PubChem CID 80556
IUPAC Name octanehydrazide
SMILES CCCCCCCC(=O)NN

Specifications

Melting Point 90°C
Color White-Yellow
Quantity 25 g
Formula Weight 158.25
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material n-Octanohydrazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.