Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Octylamine, 99+%

Catalog No. AC129491000
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Jerrycan

CAS: 111-86-4 | C8H19N | 129.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-86-4
InChI Key IOQPZZOEVPZRBK-UHFFFAOYSA-N
Synonym octylamine, n-octylamine, 1-aminooctane, 1-octanamine, 1-octylamine, caprylamine, caprylylamine, armeen 8, n-octylamine, mono, armeen 8d
PubChem CID 8143
ChEBI CHEBI:7728
IUPAC Name octan-1-amine
SMILES CCCCCCCCN

Specifications

Melting Point -1.0°C
Density 0.7820g/mL
Boiling Point 179.0°C
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Molecular Formula C8H19N
Refractive Index 1.4280 to 1.4300
Linear Formula CH3(CH2)7NH2
Quantity 100 g
Beilstein 04,196
Specific Gravity 0.782
Solubility Information Solubility in water: 0.2g/L water (25°C). Other solubilities: soluble in most common organic solvents
Molecular Weight (g/mol) 129.24
Viscosity 1.39 mPa.s (20°C)
Formula Weight 129.24
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material n-Octylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.
May cause respiratory irritation.
Very toxic to aquatic l
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee
EINECSNumber 203-916-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.