Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Palmitoyl-D-erythro-sphingosine, synthetical, 98% (99+% e.e.)

Catalog No. p-3758625
Click to view available options
Quantity:
5 mg
25 mg
250 mg
Packaging:
Glass bottle

CAS: 24696-26-2 | C34H67NO3 | 537.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24696-26-2
Molecular Formula C34H67NO3
Molecular Weight (g/mol) 537.91
MDL Number MFCD00056205
InChI Key YDNKGFDKKRUKPY-UHFFFAOYNA-N
Synonym c16-ceramide, d-erythro-c16-ceramide, n-palmitoyl 4-sphingenine, n-palmitoyl-d-sphingosine, d-erythro-4-ceramide, n-palmitoylsphingosinee, d-erythro-n-palmitoylsphingosine, n-hexadecanoyl-d-erythro-ceramide, d-erythro-n-hexadecanoylsphingenine, 2s,3r,4e-2-n-palmitoyloctadecasphinga-4-ene
PubChem CID 71314645
SMILES CCCCCCCCCCCCCCCC(=O)NC(CO)C(O)C=CCCCCCCCCCCCCC

Specifications

Melting Point 94.0°C to 95.0°C
Color White
Enantiomeric Excess 99.0% min.
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5 mg
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in chloroform,soluble in dichloromethane,insoluble in acetone
IUPAC Name N-(1,3-dihydroxyoctadec-4-en-2-yl)hexadecanamide
Formula Weight 537.91
Percent Purity 98% (99+% E.E.)
Physical Form Crystalline Powder
Chemical Name or Material N-Palmitoyl-D-erythro-sphingosine, Synthetical, ≥99% ee
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.